Assay Details
| Gene Symbol: Mapk12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013871.3 | NP_038899.1 | 3 - 4 | 550 | 76 | |
| GenBank mRNA | AK011286.1 | - | 2 - 3 | 590 | 76 | |
| AK178329.1 | - | 1 - 2 | 65 | 76 | ||
| AK178955.1 | - | 1 - 2 | 65 | 76 | ||
| AK188121.1 | - | 1 - 2 | 65 | 76 | ||
| BC021640.1 | - | 3 - 4 | 468 | 3979667 | 76 | |
| Y13439.1 | - | 3 - 4 | 486 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mapk12
Genomic Map