Assay Details
| Gene Symbol: Senp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144851.5 | NP_659100.1 | 8 - 9 | 1262 | 87 | |
| GenBank mRNA | AK028565.1 | - | 8 - 9 | 1004 | 87 | |
| AK035581.1 | - | 8 - 9 | 1098 | 87 | ||
| AK035723.1 | - | 8 - 9 | 1367 | 87 | ||
| AK054521.1 | - | 9 - 10 | 1394 | 87 | ||
| AK088597.1 | - | 8 - 9 | 1006 | 87 | ||
| AK138013.1 | - | 8 - 9 | 1063 | 87 | ||
| BC023129.1 | - | 7 - 8 | 976 | 3595534 | 87 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Senp1
Genomic Map