Assay Details
| Gene Symbol: Msl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028722.2 | NP_082998.2 | 6 - 7 | 1625 | 83 | |
| GenBank mRNA | AK014463.1 | - | 6 - 7 | 1878 | 83 | |
| AK015496.1 | - | 5 - 6 | 841 | 83 | ||
| AK053719.1 | - | 5 - 6 | 1547 | 83 | ||
| BC058629.1 | - | 6 - 7 | 1625 | 6402215 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Msl1
Genomic Map