Assay Details
| Gene Symbol: Smgc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198927.3 | NP_945121.1 | 1 - 2 | 90 | 66 | |
| GenBank mRNA | AK017450.1 | - | 1 - 2 | 91 | 66 | |
| AK132579.1 | - | 1 - 2 | 92 | 66 | ||
| AY459348.1 | - | 1 - 2 | 91 | 66 | ||
| BC055490.1 | - | 1 - 2 | 72 | 66 | ||
| BC138091.1 | - | 1 - 2 | 91 | 8861110 | 66 | |
| BC145136.1 | - | 1 - 2 | 91 | 9053684 | 66 | |
More Information
Set Membership: |
|

Smgc
Genomic Map