Assay Details
| Gene Symbol: Smgc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198927.3 | NP_945121.1 | 4 - 5 | 470 | 98 | |
| GenBank mRNA | AK017450.1 | - | 4 - 5 | 471 | 98 | |
| AK132579.1 | - | 4 - 5 | 472 | 98 | ||
| AY459348.1 | - | 4 - 5 | 471 | 98 | ||
| BC055490.1 | - | 4 - 5 | 452 | 98 | ||
| BC092073.1 | - | 3 - 4 | 291 | 98 | ||
| BC138091.1 | - | 4 - 5 | 471 | 8861110 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Smgc
Genomic Map