Assay Details
| Gene Symbol: Smgc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198927.3 | NP_945121.1 | 5 - 6 | 646 | 95 | |
| GenBank mRNA | AK132579.1 | - | 5 - 6 | 648 | 95 | |
| AY459348.1 | - | 5 - 6 | 647 | 95 | ||
| BC055490.1 | - | 5 - 6 | 628 | 95 | ||
| BC092073.1 | - | 4 - 5 | 467 | 95 | ||
| BC138091.1 | - | 5 - 6 | 647 | 8861110 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Smgc
Genomic Map