Assay Details
| Gene Symbol: Smgc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198927.3 | NP_945121.1 | 16 - 17 | 1908 | 80 | |
| GenBank mRNA | AK132579.1 | - | 15 - 16 | 1811 | 80 | |
| AY459348.1 | - | 16 - 17 | 1909 | 80 | ||
| BC022763.1 | - | 2 - 3 | 264 | 80 | ||
| BC038603.1 | - | 6 - 7 | 664 | 80 | ||
| BC049243.1 | - | 8 - 9 | 913 | 80 | ||
| BC049976.1 | - | 2 - 3 | 282 | 80 | ||
| BC055489.1 | - | 2 - 3 | 254 | 80 | ||
| BC069958.1 | - | 7 - 8 | 740 | 80 | ||
| BC138091.1 | - | 16 - 17 | 1909 | 8861110 | 80 | |
| BC145136.1 | - | 11 - 12 | 1192 | 9053684 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Smgc
Genomic Map