Assay Details
Gene Symbol: Smgc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198927.3 | NP_945121.1 | 15 - 16 | 1734 | 72 | |
GenBank mRNA | AK132579.1 | - | 14 - 15 | 1637 | 72 | |
AY459348.1 | - | 15 - 16 | 1735 | 72 | ||
BC022763.1 | - | 1 - 2 | 90 | 72 | ||
BC038603.1 | - | 5 - 6 | 490 | 72 | ||
BC049243.1 | - | 7 - 8 | 739 | 72 | ||
BC049976.1 | - | 1 - 2 | 108 | 72 | ||
BC055489.1 | - | 1 - 2 | 80 | 72 | ||
BC069958.1 | - | 6 - 7 | 566 | 72 | ||
BC138091.1 | - | 15 - 16 | 1735 | 8861110 | 72 | |
BC145136.1 | - | 10 - 11 | 1018 | 9053684 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1958720 | BC069958.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1958720 | BC092073.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1958720 | BC049243.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1958720 | BC049976.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1958720 | BC055489.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1958720 | BC069958.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1958720 | BC092073.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1958720 | BC049243.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1958720 | BC049976.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1958720 | BC055489.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30437794 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30437792 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30437790 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30437788 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30437784 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30437802 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30437799 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30437785 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30437787 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30437796 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30437803 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30437797 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30437791 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30437800 | - | - |
Whole Mouse Genome G4122A | A_51_P391560 | BC069958.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P391560 | BC092073.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P391560 | BC049243.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P391560 | BC049976.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P391560 | BC055490.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128349 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12010967 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437936 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437857 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437799 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437820 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437791 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437843 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437852 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437806 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437850 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437792 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437911 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437941 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437829 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437934 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437887 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437944 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437813 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437802 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437882 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437818 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437875 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437848 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437819 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437876 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437910 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437884 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437858 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437890 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437855 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437832 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437932 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437927 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437827 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437905 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437826 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437865 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437786 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437825 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437912 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437894 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437822 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437881 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437925 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437939 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437787 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437877 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437880 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437804 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437794 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437790 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437922 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437797 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437929 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437902 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437800 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437815 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437917 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437810 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437869 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437796 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437788 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437915 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437817 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437785 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437874 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437893 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437920 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437879 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30437886 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30437845 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30437852 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30437806 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30437822 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30437819 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30437813 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30437858 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30437794 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30437797 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30437804 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30437826 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11859092 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30437785 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30437787 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30437796 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30437857 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30437791 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30437788 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30437865 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30437810 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12340770 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30437832 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30437786 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30437811 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30437848 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30437868 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30437790 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30437869 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30437825 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30437799 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12009908 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30437850 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30437820 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12009896 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30437802 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30437827 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30437840 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30437817 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12340750 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11859095 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30437815 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30437800 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30437871 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30437829 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30437849 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10426371 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10426377 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10426375 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10426380 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10426382 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10426373 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10426372 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10426376 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10426381 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10426379 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10426378 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10426374 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1431727_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1428318_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10426370 | - | - |
Mouse Genome 430 2.0 Array | 1431727_at | - | - |
Mouse Genome 430 2.0 Array | 1428318_at | - | - |
Mouse Genome 430 2.0 Array | 1428318_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1431727_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 223809_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 223809_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1500007179.mm.1 | NM_198927.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500007179.mm.1 | NM_198927.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500007179.mm.1 | TC1500000900.mm.1 | MTA TC |
Mouse Expression Set 430 | 1431727_at | - | - |
Mouse Expression Set 430 | 1428318_PM_at | - | - |
Mouse Expression Set 430 | 1428318_at | - | - |
Mouse Expression Set 430 | 1431727_PM_at | - | - |
Murine Genome U74v2 Set | 131555_PM_at | - | - |
Murine Genome U74v2 Set | 165493_r_at | - | - |
Murine Genome U74v2 Set | 131555_at | - | - |
Murine Genome U74v2 Set | 165493_PM_r_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1431727_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1431727_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1428318_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1428318_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 796191 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 1980576 | - | - |
MouseWG-6 v2 Expression BeadChip | 1780048 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 4540435 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 1780048 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 1980576 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004540435 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001780048 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001980576 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2620853 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2602496 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2775962 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2775962 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2620853 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2602496 | - | - |
mouse-6 v1.1 (Illumina) | scl47600.16.1_2-S | - | - |
mouse-6 v1.1 (Illumina) | scl0002588.1_21-S | - | - |
mouse-6 v1.1 (Illumina) | scl0002446.1_7-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1780048 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1980576 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4540435 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0002446.1_7-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0002588.1_21-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl47600.16.1_2-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4540435 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1980576 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1780048 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2775962 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2602496 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2620853 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2602496 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2620853 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2775962 | - | - |
