Assay Details
| Gene Symbol: Ttc33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK050426.1 | - | 1 - 2 | 48 | 64 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK050426.1 transcripts.
Set Membership: |
|

Ttc33
Genomic Map