Assay Details
| Gene Symbol: Agxt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031851.1 | NP_001027021.1 | 8 - 9 | 967 | 70 | |
| GenBank mRNA | AK149538.1 | - | 8 - 9 | 894 | 70 | |
| BC141248.1 | - | 8 - 9 | 907 | 9055860 | 70 | |
| BC145576.1 | - | 8 - 9 | 904 | 9054142 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Agxt2
Genomic Map