Assay Details
| Gene Symbol: Scrn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146027.2 | NP_666139.1 | 1 - 2 | 110 | 85 | |
| GenBank mRNA | AK075869.1 | - | 1 - 2 | 74 | 85 | |
| BC010784.1 | - | 1 - 2 | 61 | 85 | ||
| BC021346.1 | - | 1 - 2 | 79 | 4163541 | 85 | |
| CN691874.1 | - | 1 - 2 | 110 | 85 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK075869.1, BC021346.1, NM_146027.2 transcripts.
Set Membership: |
|

Scrn2
Genomic Map