Assay Details
| Gene Symbol: Zfp207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011751.3 | NP_035881.1 | 7 - 8 | 836 | 85 | |
| GenBank mRNA | AB013357.1 | - | 7 - 8 | 770 | 85 | |
More Information
Set Membership: |
|

Zfp207
Genomic Map