Assay Details
| Gene Symbol: Trim25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009546.2 | NP_033572.2 | 4 - 5 | 1178 | 96 | |
| GenBank mRNA | AK154533.1 | - | 4 - 5 | 1171 | 96 | |
| AK160898.1 | - | 4 - 5 | 1130 | 96 | ||
| AK169562.1 | - | 4 - 5 | 1179 | 96 | ||
| BC006908.1 | - | 4 - 5 | 1147 | 2651664 | 96 | |
| BC034276.1 | - | 4 - 5 | 1150 | 4022161 | 96 | |
| D63902.1 | - | 4 - 5 | 1101 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trim25
Genomic Map