Assay Details
| Gene Symbol: Trim25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009546.2 | NP_033572.2 | 7 - 8 | 1365 | 68 | |
| GenBank mRNA | AK154533.1 | - | 7 - 8 | 1358 | 68 | |
| AK169562.1 | - | 7 - 8 | 1366 | 68 | ||
| AK171744.1 | - | 6 - 7 | 669 | 68 | ||
| BC006908.1 | - | 6 - 7 | 1310 | 2651664 | 68 | |
| D63902.1 | - | 7 - 8 | 1288 | 68 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Trim25
Genomic Map