Assay Details
Gene Symbol: Mbtd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134012.3 | NP_598773.2 | 4 - 5 | 657 | 75 | |
GenBank mRNA | AK141155.1 | - | 4 - 5 | 653 | 75 | |
AK153919.1 | - | 6 - 7 | 492 | 75 | ||
BC062907.1 | - | 4 - 5 | 657 | 6813849 | 75 | |
BC064014.1 | - | 6 - 7 | 761 | 6335954 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
