Assay Details
Gene Symbol: Mbtd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134012.3 | NP_598773.2 | 6 - 7 | 857 | 99 | |
GenBank mRNA | AK033238.1 | - | 1 - 2 | 3730 | 99 | |
AK141155.1 | - | 6 - 7 | 852 | 99 | ||
AK153919.1 | - | 8 - 9 | 692 | 99 | ||
BC020018.1 | - | 2 - 3 | 168 | 2646754 | 99 | |
BC062907.1 | - | 6 - 7 | 857 | 6813849 | 99 | |
BC064014.1 | - | 8 - 9 | 961 | 6335954 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
