Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100013920.mm.1 |
NM_001033186.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100013920.mm.1 |
NM_001033186.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100013920.mm.1 |
NM_001177898.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100013920.mm.1 |
NM_001177898.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100013920.mm.1 |
NM_001177899.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100013920.mm.1 |
NM_001177899.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100013920.mm.1 |
TC1100001504.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100013921.mm.1 |
NM_001033186.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100013921.mm.1 |
NM_001033186.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100013921.mm.1 |
NM_001177898.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100013921.mm.1 |
NM_001177898.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100013921.mm.1 |
NM_001177899.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100013921.mm.1 |
NM_001177899.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100013921.mm.1 |
TC1100001504.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100013919.mm.1 |
NM_001033186.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100013919.mm.1 |
NM_001033186.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100013919.mm.1 |
NM_001177898.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100013919.mm.1 |
NM_001177898.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100013919.mm.1 |
NM_001177899.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100013919.mm.1 |
NM_001177899.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100013919.mm.1 |
TC1100001504.mm.1 |
MTA TC |