Assay Details
| Gene Symbol: Mocs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113374.1 | NP_001106845.1 | 1 - 2 | 90 | 67 | |
| NM_001113375.1 | NP_001106846.1 | 1 - 2 | 90 | 67 | ||
| GenBank mRNA | AK002719.1 | - | 1 - 2 | 58 | 67 | |
| AK078782.1 | - | 1 - 2 | 58 | 67 | ||
| BC004631.1 | - | 1 - 2 | 62 | 67 | ||
| CX220128.1 | - | 1 - 2 | 40 | 67 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK002719.1, AK078782.1, NM_001113374.1, NM_001113375.1 transcripts.
Set Membership: |
|

Mocs2
Genomic Map