Assay Details
| Gene Symbol: Il31ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139299.2 | NP_647460.2 | 6 - 7 | 753 | 160 | |
| GenBank mRNA | AB083111.1 | - | 6 - 7 | 752 | 160 | |
| AF486621.1 | - | 3 - 4 | 332 | 160 | ||
| AY509150.1 | - | 649 | 160 | |||
| AY509151.1 | - | 649 | 160 | |||
| BC066164.1 | - | 4 - 5 | 474 | 30475340 | 160 | |
| BC145083.1 | - | 5 - 6 | 656 | 160 | ||
| BC145084.1 | - | 5 - 6 | 656 | 160 | ||
| BC145085.1 | - | 6 - 7 | 779 | 160 | ||
More Information
Set Membership: |
|

Il31ra
Genomic Map