Assay Details
| Gene Symbol: Hmgcs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291439.1 | NP_001278368.1 | 2 - 3 | 616 | 60 | |
| NM_145942.5 | NP_666054.2 | 3 - 4 | 675 | 60 | ||
| GenBank mRNA | AK031297.1 | - | 3 - 4 | 564 | 60 | |
| AK044835.1 | - | 3 - 4 | 565 | 60 | ||
| AK045094.1 | - | 2 - 3 | 504 | 60 | ||
| AK078743.1 | - | 2 - 3 | 506 | 60 | ||
| AK135551.1 | - | 2 - 3 | 504 | 60 | ||
| AK136171.1 | - | 3 - 4 | 564 | 60 | ||
| AK144384.1 | - | 2 - 3 | 504 | 60 | ||
| AK145424.1 | - | 2 - 3 | 504 | 60 | ||
| AK146042.1 | - | 2 - 3 | 506 | 60 | ||
| AK146590.1 | - | 2 - 3 | 506 | 60 | ||
| BC023851.1 | - | 3 - 4 | 591 | 5347038 | 60 | |
| BC029693.1 | - | 2 - 3 | 521 | 5375374 | 60 | |
| BC031363.1 | - | 2 - 3 | 523 | 5357048 | 60 | |
| BC034317.1 | - | 1 - 2 | 467 | 5366786 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hmgcs1
Genomic Map