Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200006288.mm.1 |
NM_001195767.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200006288.mm.1 |
NM_001195767.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200006288.mm.1 |
TC1200000858.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200006287.mm.1 |
NM_001195767.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200006287.mm.1 |
NM_001195767.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200006287.mm.1 |
TC1200000858.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200006286.mm.1 |
NM_001195767.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200006286.mm.1 |
NM_001195767.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200006286.mm.1 |
TC1200000858.mm.1 |
MTA TC |