Assay Details
Gene Symbol: Map3k1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011945.2 | NP_036075.2 | 10 - 11 | 1968 | 77 | |
GenBank mRNA | AF117340.1 | - | 10 - 11 | 1968 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
