Assay Details
| Gene Symbol: Mtap7d3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177293.3 | NP_796267.2 | 10 - 11 | 1745 | 130 | |
| GenBank mRNA | AK030211.1 | - | 9 - 10 | 1662 | 130 | |
| AK048385.1 | - | 4 - 5 | 755 | 130 | ||
| BC096764.1 | - | 10 - 11 | 1747 | 130 | ||
| BC118963.2 | - | 9 - 10 | 1662 | 40095274 | 130 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mtap7d3
Genomic Map