Assay Details
Gene Symbol: Mtap7d3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177293.3 | NP_796267.2 | 2 - 3 | 176 | 76 | |
GenBank mRNA | AK030211.1 | - | 1 - 2 | 93 | 76 | |
AK049164.1 | - | 2 - 3 | 166 | 76 | ||
BB610806.1 | - | 2 - 3 | 177 | 76 | ||
BC096764.1 | - | 2 - 3 | 177 | 76 | ||
BC118963.2 | - | 1 - 2 | 93 | 40095274 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
