Assay Details
| Gene Symbol: Mtap7d3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177293.3 | NP_796267.2 | 3 - 4 | 258 | 71 | |
| GenBank mRNA | AK030211.1 | - | 2 - 3 | 175 | 71 | |
| AK049164.1 | - | 3 - 4 | 248 | 71 | ||
| BB610806.1 | - | 3 - 4 | 259 | 71 | ||
| BC096764.1 | - | 3 - 4 | 259 | 71 | ||
| BC118963.2 | - | 2 - 3 | 175 | 40095274 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mtap7d3
Genomic Map