Assay Details
| Gene Symbol: Mtap7d3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177293.3 | NP_796267.2 | 8 - 9 | 1444 | 64 | |
| GenBank mRNA | AK030211.1 | - | 7 - 8 | 1361 | 64 | |
| AK048385.1 | - | 2 - 3 | 454 | 64 | ||
| AK049164.1 | - | 8 - 9 | 1344 | 64 | ||
| BC096764.1 | - | 8 - 9 | 1446 | 64 | ||
| BC118963.2 | - | 7 - 8 | 1361 | 40095274 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mtap7d3
Genomic Map