Assay Details
| Gene Symbol: Mtap7d3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177293.3 | NP_796267.2 | 1 - 2 | 91 | 76 | |
| GenBank mRNA | AK049164.1 | - | 1 - 2 | 81 | 76 | |
| BB610806.1 | - | 1 - 2 | 92 | 76 | ||
| BC096764.1 | - | 1 - 2 | 92 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mtap7d3
Genomic Map