Assay Details
Gene Symbol: 1190003K10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195435.1 | NP_001182364.1 | 2 - 3 | 487 | 91 | |
GenBank mRNA | AK004471.1 | - | 2 - 3 | 488 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2037892 | NM_001195435.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2037892 | AK004471.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 121780 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 174816 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 796638 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 612046 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2037892 | NM_001195435.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2037892 | AK004471.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02709907 | - | - |
Whole Mouse Genome G4122A | A_51_P337015 | NM_001195435.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P337015 | AK004471.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139648 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10918616 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29099034 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29099052 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29099141 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29099037 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29099056 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29099039 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29099030 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29099154 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29099054 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29099062 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29099158 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29099152 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29099040 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29099057 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29099027 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29099147 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29099060 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29099156 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29099019 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10626380 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29099155 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29099143 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29099041 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29099045 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29099049 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29099032 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29099024 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29099020 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10899519 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29099159 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29099021 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29099035 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29099149 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29099029 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29099044 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29099023 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29099038 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT MG-430 PM Array Plate | 1432300_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1432300_at | - | - |
Mouse Genome 430 2.0 Array | 1432300_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68880_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68880_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1300014239.mm.1 | NM_001195435.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300014239.mm.1 | NM_001195435.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300014239.mm.1 | TC1300002206.mm.1 | MTA TC |
Mouse Expression Set 430 | 1432300_at | - | - |
Mouse Expression Set 430 | 1432300_PM_at | - | - |
Murine Genome U74v2 Set | 170886_at | - | - |
Murine Genome U74v2 Set | 170886_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1432300_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1432300_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 475923 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.1 expression beadchip | ILMN_2493688 | - | - |
