Assay Details
| Gene Symbol: Srek1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172592.2 | NP_766180.1 | 6 - 7 | 1410 | 71 | |
| GenBank mRNA | AK028794.1 | - | 2 - 3 | 2946 | 71 | |
| AK033354.1 | - | 6 - 7 | 1410 | 71 | ||
| AK037115.1 | - | 6 - 7 | 1386 | 71 | ||
| AK039311.1 | - | 8 - 9 | 1145 | 71 | ||
| AK044086.1 | - | 5 - 6 | 714 | 71 | ||
| AK052265.1 | - | 8 - 9 | 1279 | 71 | ||
| AK054414.1 | - | 6 - 7 | 2049 | 71 | ||
| BC063761.1 | - | 9 - 10 | 1155 | 71 | ||
| BC070460.1 | - | 6 - 7 | 783 | 30548562 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Srek1
Genomic Map