Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200005994.mm.1 |
NM_030225.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200005994.mm.1 |
NM_030225.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200005994.mm.1 |
TC1200000828.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200006000.mm.1 |
NM_030225.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200006000.mm.1 |
NM_030225.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200006000.mm.1 |
TC1200000828.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200005996.mm.1 |
NM_030225.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200005996.mm.1 |
NM_030225.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200005996.mm.1 |
TC1200000828.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200005998.mm.1 |
NM_030225.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200005998.mm.1 |
NM_030225.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200005998.mm.1 |
TC1200000828.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200005995.mm.1 |
NM_030225.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200005995.mm.1 |
NM_030225.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200005995.mm.1 |
TC1200000828.mm.1 |
MTA TC |