Assay Details
Gene Symbol: Dlst | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030225.4 | NP_084501.1 | 5 - 6 | 340 | 75 | |
GenBank mRNA | AK019713.2 | - | 337 | 75 | ||
AK054053.1 | - | 5 - 6 | 294 | 75 | ||
AK149664.1 | - | 5 - 6 | 316 | 75 | ||
AK158877.1 | - | 5 - 6 | 341 | 75 | ||
AK168570.1 | - | 5 - 6 | 316 | 75 | ||
AK169943.1 | - | 5 - 6 | 294 | 75 | ||
BC006702.1 | - | 5 - 6 | 277 | 3583936 | 75 | |
CT010197.1 | - | 5 - 6 | 276 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
