Assay Details
Gene Symbol: 1700125H20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK007281.1 | - | 2 - 3 | 133 | 100 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK007281.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
