Assay Details
Gene Symbol: Abi3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163464.1 | NP_001156936.1 | 4 - 5 | 497 | 100 | |
NM_025659.4 | NP_079935.1 | 4 - 5 | 848 | 100 | ||
GenBank mRNA | AK008928.1 | - | 4 - 5 | 570 | 100 | |
AK018381.1 | - | 4 - 5 | 569 | 100 | ||
AK033965.1 | - | 4 - 5 | 569 | 100 | ||
AK037144.1 | - | 4 - 5 | 498 | 100 | ||
AK172098.1 | - | 4 - 5 | 567 | 100 | ||
BC058260.1 | - | 4 - 5 | 558 | 6474610 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
