Assay Details
Gene Symbol: Med6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027213.2 | NP_081489.2 | 6 - 7 | 577 | 89 | |
GenBank mRNA | AK005232.1 | - | 6 - 7 | 622 | 89 | |
AK171784.1 | - | 6 - 7 | 593 | 89 | ||
BC013096.1 | - | 6 - 7 | 577 | 3710612 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
