Assay Details
| Gene Symbol: Cdh7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172853.2 | NP_766441.1 | 3 - 4 | 851 | 70 | |
| GenBank mRNA | AK034096.1 | - | 3 - 4 | 851 | 70 | |
| AK042657.1 | - | 5 - 6 | 1246 | 70 | ||
| AK137369.1 | - | 3 - 4 | 774 | 70 | ||
| BC083189.1 | - | 4 - 5 | 1441 | 70 | ||
More Information
Set Membership: |
|

Cdh7
Genomic Map