Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500018495.mm.1 |
NM_001161767.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018495.mm.1 |
NM_001161767.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018495.mm.1 |
NM_001161768.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018495.mm.1 |
NM_001161768.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018495.mm.1 |
NM_172451.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018495.mm.1 |
NM_172451.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018495.mm.1 |
TC1500002250.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500018493.mm.1 |
NM_001161767.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018493.mm.1 |
NM_001161767.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018493.mm.1 |
NM_001161768.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018493.mm.1 |
NM_001161768.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018493.mm.1 |
NM_172451.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018493.mm.1 |
NM_172451.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018493.mm.1 |
TC1500002250.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500018494.mm.1 |
NM_001161767.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018494.mm.1 |
NM_001161767.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018494.mm.1 |
NM_001161768.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018494.mm.1 |
NM_001161768.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018494.mm.1 |
NM_172451.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018494.mm.1 |
NM_172451.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018494.mm.1 |
TC1500002250.mm.1 |
MTA TC |