Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500018594.mm.1 |
NM_008474.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018594.mm.1 |
NM_008474.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018594.mm.1 |
TC1500002261.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500018595.mm.1 |
NM_008474.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018595.mm.1 |
NM_008474.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018595.mm.1 |
TC1500002261.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500018596.mm.1 |
NM_008474.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018596.mm.1 |
NM_008474.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018596.mm.1 |
TC1500002261.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500018593.mm.1 |
NM_008474.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018593.mm.1 |
NM_008474.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018593.mm.1 |
TC1500002261.mm.1 |
MTA TC |