Assay Details
Gene Symbol: Krt84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008474.2 | NP_032500.2 | 5 - 6 | 1173 | 87 | |
GenBank mRNA | AY028607.1 | - | 5 - 6 | 1130 | 87 | |
BC114971.1 | - | 5 - 6 | 1126 | 40039508 | 87 | |
X65505.1 | - | 2 - 3 | 193 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
