Assay Details
Gene Symbol: Krt75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133357.3 | NP_579935.1 | 6 - 7 | 1239 | 81 | |
GenBank mRNA | AF343088.1 | - | 5 - 6 | 657 | 81 | |
AK028972.1 | - | 6 - 7 | 1240 | 81 | ||
AK037233.1 | - | 6 - 7 | 1240 | 81 | ||
AK076367.1 | - | 6 - 7 | 1240 | 81 | ||
BC137935.1 | - | 6 - 7 | 1240 | 8860954 | 81 | |
BC137936.1 | - | 6 - 7 | 1240 | 8860955 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
