Assay Details
Gene Symbol: Krt24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029393.1 | NP_083669.1 | 6 - 7 | 1384 | 89 | |
GenBank mRNA | AK009986.1 | - | 5 - 6 | 729 | 89 | |
AK010165.1 | - | 6 - 7 | 1407 | 89 | ||
AK053678.1 | - | 6 - 7 | 1130 | 89 | ||
BC129847.1 | - | 6 - 7 | 1384 | 40107361 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
