Assay Details
Gene Symbol: Krt24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029393.1 | NP_083669.1 | 7 - 8 | 1466 | 136 | |
GenBank mRNA | AK009986.1 | - | 6 - 7 | 811 | 136 | |
AK010165.1 | - | 7 - 8 | 1489 | 136 | ||
AK053678.1 | - | 7 - 8 | 1212 | 136 | ||
BC129847.1 | - | 7 - 8 | 1466 | 40107361 | 136 | |
BC129848.1 | - | 7 - 8 | 1466 | 40107363 | 136 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P104945 | AK053678.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P104945 | BC129847.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P104945 | BC129848.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P104945 | GU046702.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P104945 | NM_029393.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 902119 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 308112 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 44253 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 118961 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P119042 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P107626 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P144532 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P104945 | AK053678.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P104945 | BC129847.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P104945 | BC129848.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P104945 | GU046702.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P104945 | NM_029393.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P107626 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P119042 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P144532 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P107626 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P144532 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P119042 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061531 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061465 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061542 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061524 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061472 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061517 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061441 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061537 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061545 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061477 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061481 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061520 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061507 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061427 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061540 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061552 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061468 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061496 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061480 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061509 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061501 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061503 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061448 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061492 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061432 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061474 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061551 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061533 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061423 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061424 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061519 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061434 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061454 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061457 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061494 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061449 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061512 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061455 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061515 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061489 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061445 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061488 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061460 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061450 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061467 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061513 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061483 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061446 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061502 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061443 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061527 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061438 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061510 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061525 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061544 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061498 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09128380 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061496 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09060689 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061492 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09255157 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061549 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09259755 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061525 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061502 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09255164 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061537 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061480 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061542 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061513 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061489 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061531 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061512 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061503 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09598845 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061509 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061510 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09496401 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09128379 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061477 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061468 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061527 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061463 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061486 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061465 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061481 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061488 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09364022 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09598840 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061472 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061507 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09490280 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061517 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061524 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061501 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061534 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061533 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061474 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061519 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061470 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061483 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061538 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28061467 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10390790 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10390794 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10390791 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10390797 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10390792 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10390795 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10390793 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10390796 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1453327_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10390789 | - | - |
Mouse Genome 430 2.0 Array | 1453327_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1453327_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75706_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75706_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1100034137.mm.1 | NM_029393.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100034137.mm.1 | NM_029393.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100034137.mm.1 | TC1100003700.mm.1 | MTA TC |
Mouse Expression Set 430 | 1453327_at | - | - |
Mouse Expression Set 430 | 1453327_PM_at | - | - |
Murine Genome U74v2 Set | 165952_PM_r_at | - | - |
Murine Genome U74v2 Set | 165952_r_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1453327_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1453327_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 567553 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1243795 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1243795 | - | - |
