Assay Details
| Gene Symbol: Bcas3os1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045875.1 | 1 - 2 | 41 | 72 | ||
| GenBank mRNA | AK053124.1 | - | 1 - 2 | 42 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bcas3os1
Genomic Map