Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1300018499.mm.1 |
NR_045508.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300018499.mm.1 |
NR_045508.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300018499.mm.1 |
TC1300002733.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300018496.mm.1 |
NR_045508.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300018496.mm.1 |
NR_045508.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300018496.mm.1 |
TC1300002733.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300018497.mm.1 |
NR_045508.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300018497.mm.1 |
NR_045508.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300018497.mm.1 |
TC1300002733.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300018501.mm.1 |
NR_045508.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300018501.mm.1 |
NR_045508.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300018501.mm.1 |
TC1300002733.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300018504.mm.1 |
NR_045508.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300018504.mm.1 |
NR_045508.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300018504.mm.1 |
TC1300002733.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300018494.mm.1 |
NR_045508.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300018494.mm.1 |
NR_045508.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300018494.mm.1 |
TC1300002733.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300018495.mm.1 |
NR_045508.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300018495.mm.1 |
NR_045508.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300018495.mm.1 |
TC1300002733.mm.1 |
MTA TC |