Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700002347.mm.1 |
NM_175229.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700002347.mm.1 |
NM_175229.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700002347.mm.1 |
TC1700000323.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700002350.mm.1 |
NM_175229.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700002350.mm.1 |
NM_175229.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700002350.mm.1 |
TC1700000323.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700002349.mm.1 |
NM_175229.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700002349.mm.1 |
NM_175229.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700002349.mm.1 |
TC1700000323.mm.1 |
MTA TC |