Assay Details
Gene Symbol: Zranb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285945.1 | NP_001272874.1 | 12 - 13 | 1618 | 133 | |
NM_027678.3 | NP_081954.1 | 12 - 13 | 1757 | 133 | ||
GenBank mRNA | AK050039.1 | - | 8 - 9 | 1038 | 133 | |
AK083641.1 | - | 12 - 13 | 1736 | 133 | ||
AK084943.1 | - | 12 - 13 | 1741 | 133 | ||
BC066035.1 | - | 12 - 13 | 1694 | 6837116 | 133 | |
BC117921.1 | - | 12 - 13 | 1572 | 40097463 | 133 | |
BC117922.2 | - | 12 - 13 | 2764 | 40097464 | 133 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
