Assay Details
| Gene Symbol: Zranb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285945.1 | NP_001272874.1 | 13 - 14 | 2027 | 96 | |
| NM_027678.3 | NP_081954.1 | 13 - 14 | 2166 | 96 | ||
| GenBank mRNA | AK050039.1 | - | 9 - 10 | 1447 | 96 | |
| AK083641.1 | - | 13 - 14 | 2145 | 96 | ||
| BC066035.1 | - | 13 - 14 | 2103 | 6837116 | 96 | |
| BC117921.1 | - | 13 - 14 | 1981 | 40097463 | 96 | |
| BC117922.2 | - | 13 - 14 | 3173 | 40097464 | 96 | |
More Information
Set Membership: |
|

Zranb3
Genomic Map