Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700015645.mm.1 |
NM_001162868.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700015645.mm.1 |
NM_001162868.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700015645.mm.1 |
NM_001162869.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700015645.mm.1 |
NM_001162869.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700015645.mm.1 |
NM_153140.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700015645.mm.1 |
NM_153140.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700015645.mm.1 |
TC1700001686.mm.1 |
MTA TC |