Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700015720.mm.1 |
NM_001081070.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700015720.mm.1 |
NM_001081070.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700015720.mm.1 |
TC1700002806.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700015723.mm.1 |
NM_001081070.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700015723.mm.1 |
NM_001081070.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700015723.mm.1 |
TC1700002806.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700015721.mm.1 |
NM_001081070.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700015721.mm.1 |
NM_001081070.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700015721.mm.1 |
TC1700002806.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700015722.mm.1 |
NM_001081070.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700015722.mm.1 |
NM_001081070.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700015722.mm.1 |
TC1700002806.mm.1 |
MTA TC |