Assay Details
| Gene Symbol: Pdia2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081070.1 | NP_001074539.1 | 7 - 8 | 1183 | 117 | |
| GenBank mRNA | AK007743.1 | - | 7 - 8 | 1132 | 117 | |
| BC116671.1 | - | 7 - 8 | 1149 | 40107256 | 117 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pdia2
Genomic Map