Assay Details
Gene Symbol: 1810013A23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045427.1 | 1 - 2 | 177 | 60 | ||
GenBank mRNA | AK007468.1 | - | 1 - 2 | 178 | 60 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | differentially expressed in FDCP 6 | Def6 |
2410003F05Rik, 6430538D02Rik, AV094905, Ibp, Slat, Slat2, Slat6 |
-43.1kb | Sense | Chr. 17 - 28207778 - 28228608 on NCBI Build GRCm38 |
Downstream |
Fanconi anemia, complementation group E |
Fance |
2810451D06Rik, AI415634, AW209126 |
41.3kb | Sense | Chr. 17 - 28313530 - 28326574 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
